Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack for 32/8 Flipper JR Reversible Multi-Boxes, 3 x 6 Configuration, 18 Box Capacity
SDP
Catalog No. 503029063
Encompass_Preferred
Easily access boxes from either side of rack with right or left handed doors. Our freezer racks have been configured to fit all major ultra low (-80 deg) freezers on the market; racks are designed for ultra low upright freezers. Made of corrosion resistant stainless steel Dimensions (DxWxH): 11 x 6 15/16 x 11 inches (280 x 177 x 280mm)Compartment Dimensions: 3 7/16 x 6 7/8 x 2 3/16 inches (87 x 174 x 55mm)
Catalog No. 50-302-9063 Supplier Alkali Scientific Supplier No. UFF36
Easily access boxes from either side of rack with right or left handed doors. Our freezer racks have been configured to fit all major ultra low (-80 deg) freezers on the market; racks are designed for ultra low upright freezers. Made of corrosion resistant stainless steel Dimensions (DxWxH): 11 x 6 15/16 x 11 inches (280 x 177 x 280mm)Compartment Dimensions: 3 7/16 x 6 7/8 x 2 3/16 inches (87 x 174 x 55mm)