Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack for 50-Cell 1.5ml Microtube Storage Boxes, 3x6x2 Configuration, 18 Box Capacity
SDP
Catalog No. 503027805
Encompass_Preferred
For 50-Cell 1.5 mL Storage Boxes The UFM-3615 Upright Freezer Racks are designed for storage of 50-Cell 1.5ml Microtube Boxes and feature a 3 Boxes High x 6 Boxes Deep configuration. These upright freezer racks include handles, are constructed of corrosion resistant stainless steel and offer convenient storage of samples in an organized format.  Each rack compartment holds two 50 Cell 1.5ml Microtube Storage Boxes. Microtube boxes are not included with purchase. Security devices are available for each rack though ordered separately. Dimensions (DxWxH): 19 3/16 x 5 7/8 x 12 5/8 inches (488 x 149 x 321mm)Compartment Dimensions: 6 1/8 x 5 13/16 x 2 5/16 inches (156 x 147 x 57.6mm)
Catalog No. 50-302-7805 Supplier Alkali Scientific Supplier No. UFM3615
For 50-Cell 1.5 mL Storage Boxes The UFM-3615 Upright Freezer Racks are designed for storage of 50-Cell 1.5ml Microtube Boxes and feature a 3 Boxes High x 6 Boxes Deep configuration. These upright freezer racks include handles, are constructed of corrosion resistant stainless steel and offer convenient storage of samples in an organized format.  Each rack compartment holds two 50 Cell 1.5ml Microtube Storage Boxes. Microtube boxes are not included with purchase. Security devices are available for each rack though ordered separately. Dimensions (DxWxH): 19 3/16 x 5 7/8 x 12 5/8 inches (488 x 149 x 321mm)Compartment Dimensions: 6 1/8 x 5 13/16 x 2 5/16 inches (156 x 147 x 57.6mm)