Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack for 50-Cell 0.5 ml Microtube Storage Boxes, 1 x 4 x 2 Configuration, 8 Box Capacity
SDP
Supplier:  Alkali Scientific UFM1405
Encompass_Preferred
For 50-cell 0.5mL Storage Boxes Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Durable stainless steel freezer racks facilitate organization and maximize valuable storage space.. Handles at both ends accommodate freezers with right or left handed doors. Upright Freezer Racks for 50-Cell 0.5ml Microtube Storage Boxes. Each rack compartment holds two 50 Cell 0.5ml Microtube Storage Boxes. Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Features: Holds 50-Cell 0.5ml Microtube Storage Boxes Constructed of corrosion resistant stainless steel Sturdy handle design Dimensions (DxWxH): 6 x 5 1/2 x 8 13/16 inches (153 x 140 x 224mm)Compartment Dimensions: 5 3/8 x 5 7/16 x 2 3/16 inches (137 x 138 x 55mm)
Catalog No. 50-302-8960