Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack For 96-Well and 384-Well Microtiter Plates
SDP
Catalog No. 503027794
Encompass_Preferred
Plate Capacity: 72 with lids 96 without lids6×4 Configuration Dimensions (DxWxH): 21 3/4 x 5 7/16 x 9 3/4 inches (552 x 138 x 247mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm)     *Lock device is ordered separately.
Catalog No. 50-302-7794 Supplier Alkali Scientific Supplier No. UFDMP64
Plate Capacity: 72 with lids 96 without lids6×4 Configuration Dimensions (DxWxH): 21 3/4 x 5 7/16 x 9 3/4 inches (552 x 138 x 247mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm)     *Lock device is ordered separately.