Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack for Mixed Storage of 2″,3″ and 3.75″ Standard Boxes
SDP
Catalog No. 503028033
Encompass_Preferred
For Mixed Storage of  2″,3″ and 3.75″ Standard Boxes Dimensions L x H x W: 16 13/16 x 8 9/16 x 51 1/2 inch( 427 x 218 x 140mm)Compartment Dimensions: 5 3/8 x 8 3/8 x 5 7/16 inch ( 136.5 x 213 x 138mm)
Catalog No. 50-302-8033 Supplier Alkali Scientific Supplier No. UFR308
For Mixed Storage of  2″,3″ and 3.75″ Standard Boxes Dimensions L x H x W: 16 13/16 x 8 9/16 x 51 1/2 inch( 427 x 218 x 140mm)Compartment Dimensions: 5 3/8 x 8 3/8 x 5 7/16 inch ( 136.5 x 213 x 138mm)