Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack for Mixes Storage of 2", 3", and 3.75" boxes.
SDP
Catalog No. 503027846
Encompass_Preferred
For Mixed Storage of  2″,3″ and 3.75″ Standard Boxes Dimensions L x H x W: 16 13/16 x 12 3/8 x 51 1/2 inch( 427 x 315 x 140mm)Compartment Dimensions: 5 3/8 x 12 3/16 x 5 7/16 inch ( 136.5 x 310 x 138mm)
Catalog No. 50-302-7846 Supplier Alkali Scientific Supplier No. UFR312L
For Mixed Storage of  2″,3″ and 3.75″ Standard Boxes Dimensions L x H x W: 16 13/16 x 12 3/8 x 51 1/2 inch( 427 x 315 x 140mm)Compartment Dimensions: 5 3/8 x 12 3/16 x 5 7/16 inch ( 136.5 x 310 x 138mm)