Learn More
Labrepco Inc LabRepCo UFMP-612L Upright Freezer Rack, Corrosion-Resistant Steel, Holds 108 Plates, 22 1/2 in
SDP
Catalog No. NC3364766
Encompass_Preferred
The LabRepCo upright freezer rack is designed for secure, organized storage of 96-well and 384-well microtiter plates. Constructed from high-quality stainless steel, it excels in ultra-low temperature environments, maximizing freezer space while allowing easy access to plates. It accommodates up to 108 plates with lids or 132 without and features locking rods for secure plate retention, making it an efficient solution for laboratory sample management.
Catalog No. NC3364766 Supplier Labrepco Inc Supplier No. UFMP612L
The LabRepCo upright freezer rack is designed for secure, organized storage of 96-well and 384-well microtiter plates. Constructed from high-quality stainless steel, it excels in ultra-low temperature environments, maximizing freezer space while allowing easy access to plates. It accommodates up to 108 plates with lids or 132 without and features locking rods for secure plate retention, making it an efficient solution for laboratory sample management.