Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack for SBS formatted boxes, 4x4 configuration
SDP
Catalog No. 503028449
Encompass_Preferred
Combination Drawer Racks conveniently store both boxes and centrifuge tubes in a single rack.  Constructed of corrosion resistant stainsteel. Drawer Racks facilitate both access to and storage of samples without removing the whole racks from the freezer.  Handles at both ends accommodate freezers with right or left handed doors.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Features: Combination Drawer Racks conveniently store both boxes and centrifuge tubes in a single rack Constructed of corrosion resistant stainless steel Easy drawer access to each tube and no need for boxes Security lock devices available for each rack Dimensions L x H x W: 14 9/16 x 10 11/16 x 5 7/16 Inch (370 x 136 x 138mm)Compartment Size l x h x w: 3 1/2 x 2 3/8 z 5 1/8 Inch (89 x 61 x 130mm)
Catalog No. 50-302-8449 Supplier Alkali Scientific Supplier No. UFDMM44
Combination Drawer Racks conveniently store both boxes and centrifuge tubes in a single rack.  Constructed of corrosion resistant stainsteel. Drawer Racks facilitate both access to and storage of samples without removing the whole racks from the freezer.  Handles at both ends accommodate freezers with right or left handed doors.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Features: Combination Drawer Racks conveniently store both boxes and centrifuge tubes in a single rack Constructed of corrosion resistant stainless steel Easy drawer access to each tube and no need for boxes Security lock devices available for each rack Dimensions L x H x W: 14 9/16 x 10 11/16 x 5 7/16 Inch (370 x 136 x 138mm)Compartment Size l x h x w: 3 1/2 x 2 3/8 z 5 1/8 Inch (89 x 61 x 130mm)