Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack for SBS Formatted Boxes (Matrix, Micronic, Fluidx, Abgene), 6x3 configuration, 18 Box Capacity
SDP
Catalog No. 503027985
Encompass_Preferred
For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene)- H: 15/16″- 1 1/8″ (23.8-28.45mm) Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers.  Removable clip can be adjusted so that the rack can hold both standard 6" and 3" boxes.Constructed of corrosion resistant stainless steel. Features: Modifiable Includes Handles Constructed of corrosion resistant stainless steel Dimensions (DxWxH): 21 3/4 x 5 7/16 x 3 7/8 inches (552 x 138 x 98mm)Compartment Dimensions: 3 7/16 x 5 7/16 x 1 3/16 inches (88 x 130 x 30mm)
Catalog No. 50-302-7985 Supplier Alkali Scientific Supplier No. UFMM63
For Speciality Tube Racks (Matrix, Micronic, Fluidx, Abgene)- H: 15/16″- 1 1/8″ (23.8-28.45mm) Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers.  Removable clip can be adjusted so that the rack can hold both standard 6" and 3" boxes.Constructed of corrosion resistant stainless steel. Features: Modifiable Includes Handles Constructed of corrosion resistant stainless steel Dimensions (DxWxH): 21 3/4 x 5 7/16 x 3 7/8 inches (552 x 138 x 98mm)Compartment Dimensions: 3 7/16 x 5 7/16 x 1 3/16 inches (88 x 130 x 30mm)