Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack for SBS formatted boxes, 6x4 configuration
SDP
Catalog No. 503028139
Encompass_Preferred
For Speciality Tube Racks (Matrix,Micronic,Fluidx, Abgene )- H: 15/16″-1 1/8″(23.8-28.45mm) Dimensions L x H x W: 21 5/8 x 10 11/16 x 5 7/16 Inch (549.2 x 271 x 138mm)Compartment Size l x h x w: 3 1/2 x 2 3/8 z 5 1/8 Inch (89 x 61 x 130mm)
Catalog No. 50-302-8139 Supplier Alkali Scientific Supplier No. UFDMM64
For Speciality Tube Racks (Matrix,Micronic,Fluidx, Abgene )- H: 15/16″-1 1/8″(23.8-28.45mm) Dimensions L x H x W: 21 5/8 x 10 11/16 x 5 7/16 Inch (549.2 x 271 x 138mm)Compartment Size l x h x w: 3 1/2 x 2 3/8 z 5 1/8 Inch (89 x 61 x 130mm)