Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack for Standard 2″ Boxes- Rack Only (2 Boxes Deep x 5 Boxes High)
SDP
Catalog No. 503028088
Encompass_Preferred
For Standard 2″ Boxes Standard upright freezer racks for storage of 2 inch boxes, features a 2 boxes deep by 5 boxes high configuration for a total storage of 10 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. This freezer rack does not come standard with storage boxes. Dimensions (DxHxW): 11 1/2 x 11 x 5 1/2 inches (292 x 280 x 140mm)Compartment Dimensions(DxHxW): 5 3/8 x 2 3/16 x 5 7/16 inches (136 x 55 x 138mm)
Catalog No. 50-302-8088 Supplier Alkali Scientific Supplier No. UFR252
For Standard 2″ Boxes Standard upright freezer racks for storage of 2 inch boxes, features a 2 boxes deep by 5 boxes high configuration for a total storage of 10 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. This freezer rack does not come standard with storage boxes. Dimensions (DxHxW): 11 1/2 x 11 x 5 1/2 inches (292 x 280 x 140mm)Compartment Dimensions(DxHxW): 5 3/8 x 2 3/16 x 5 7/16 inches (136 x 55 x 138mm)