Learn More
Labrepco Inc UPRIGHT FREEZER RACK FOR STD
SDP
Supplier:  Labrepco Inc UF443
Encompass_Preferred
Rack, Upright freezer; Labrepco; Crystal; For standard 3 in. boxes; Capacity (Box): 16; L x H x D: 562 x 321 x 140mm
Catalog No. 50-902-0177