Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Side Access Freezer Rack, Holds (18) Large 2" Boxes, Rack Dims 19 3/16L x 13 13/16H x 5 7/8W" (488 x 351 x 149mm) with Compartment Dims 6 1/8L x 2 5/16H x 5 13/16W" (156 x 57.6 x 147.5mm) 3x6
Catalog No. NC2208077
Encompass_Preferred
Rack Designed to hold (18) Large 2" Cardboard Boxes (2"H x5 3/4W x5 3/4L" (51 x 146 x 146mm), or 100-Cell Hinged Top Plastic Boxes, (2"H x 5 1/2W x 6L", 54 x 141 x 152mm), made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available . Security lock devices available for each rack.
This item is not returnable. View return policy
Catalog No. NC2208077 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFHT36
Rack Designed to hold (18) Large 2" Cardboard Boxes (2"H x5 3/4W x5 3/4L" (51 x 146 x 146mm), or 100-Cell Hinged Top Plastic Boxes, (2"H x 5 1/2W x 6L", 54 x 141 x 152mm), made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available . Security lock devices available for each rack.