Learn More
Labrepco Inc Value Series Upright Freezer Rack for Standard 2″ Boxes- Rack Only (4 Boxes Deep x 4 Boxes High)
SDP
Catalog No. NC1671573
Encompass_Preferred
These racks will fit the Revco ULT-17,21,25 cu.ft units, Revco EXF-320 and 400, HeraFreeze HFU-320 and 400, Harris SLT/DLT/HLT-17, 21, 25V, So-Low U85-18, 22 and 25 cu.ft units, which all have a 4 deep, 4 tall configuration. These will also fit VWR, New Brunswick and Norlake ultra low temp freezers.
Catalog No. NC1671573 Supplier Labrepco Inc Supplier No. CUF442
These racks will fit the Revco ULT-17,21,25 cu.ft units, Revco EXF-320 and 400, HeraFreeze HFU-320 and 400, Harris SLT/DLT/HLT-17, 21, 25V, So-Low U85-18, 22 and 25 cu.ft units, which all have a 4 deep, 4 tall configuration. These will also fit VWR, New Brunswick and Norlake ultra low temp freezers.