Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack (UFDMP-74) For 96-Well and 384-Well Microtiter Plates, Holds 84 with Lids, 112 without Lids
SDP
Catalog No. 503027977
Encompass_Preferred
Our (UFDMP-74) Freezer Racks are constructed of high-quality, corrosion resistant stainless steel. Durable stainless steel freezer racks facilitate organization and maximize valuable storage space..  Handles at both ends accommodate freezers with right or left handed doors.  Racks are for 96-well & 384-well Microtiter Plates.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers.   Features: Racks are for 96-well & 384-well Microtiter Plates Constructed of corrosion resistant stainless steel Sturdy handle design Plate Capacity: 84 with lids 112 without lids7×4 Configuration Dimensions (DxWxH): 25 1/4 x 5 7/16 x 9 3/4 inches (642 x 138 x 247mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm) *Lock device is ordered separately.
Catalog No. 50-302-7977 Supplier Alkali Scientific Supplier No. UFDMP74
Our (UFDMP-74) Freezer Racks are constructed of high-quality, corrosion resistant stainless steel. Durable stainless steel freezer racks facilitate organization and maximize valuable storage space..  Handles at both ends accommodate freezers with right or left handed doors.  Racks are for 96-well & 384-well Microtiter Plates.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers.   Features: Racks are for 96-well & 384-well Microtiter Plates Constructed of corrosion resistant stainless steel Sturdy handle design Plate Capacity: 84 with lids 112 without lids7×4 Configuration Dimensions (DxWxH): 25 1/4 x 5 7/16 x 9 3/4 inches (642 x 138 x 247mm)Compartment Dimensions: 3 1/2 x 5 1/8 x 2 3/16 inches (89 x 130 x 55.8mm) *Lock device is ordered separately.