Learn More
Labrepco Inc UPRIGHT FREEZER RACK W/DRAWERS
SDP
Catalog No. NC1984365
Encompass_Preferred
Value Series Upright Freezer Rack with Drawers for Standard 2″ Boxes- Rack Only (5 Boxes Deep x 4 Boxes High)Dimensions(LxHxW): 27.54" x 9.12" x 5.49"Capacity: 20 Boxes Per RackShipping Weight (lb): 12lbsIncludes: Rack Only
Catalog No. NC1984365 Supplier Labrepco Inc Supplier No. CUFT542
Value Series Upright Freezer Rack with Drawers for Standard 2″ Boxes- Rack Only (5 Boxes Deep x 4 Boxes High)Dimensions(LxHxW): 27.54" x 9.12" x 5.49"Capacity: 20 Boxes Per RackShipping Weight (lb): 12lbsIncludes: Rack Only