Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack with Drawers for Cardboard 2″ Boxes; PHCbi Upright Freezer; Rack Only; 3x6 Configuration, 18 Box Capacity
SDP
Catalog No. 503027997
Encompass_Preferred
For Standard 2″ BoxesRack Only Upright freezer racks for storage of 2 inch boxes, features a 5 boxes deep by 5 boxes high configuration for a total storage of 25 boxes per rack. These upright freezer drawer racks are built with durable, corrosion resistant stainless steel. Each rack includes separate drawers with handles allowing for an easy retrieval from a laboratory freezer without the need to pull out the entire rack. This drawer style freezer rack does not come standard with storage boxes. This rack is compatible with the PHCbi VIP freezers. Please see theFreezer Rack Configurator if you are unsure of what rack you may need. Dimensions (D x H x W): 16 7/8 x 12 5/8 x 5 1/2  inches (428 x 321 x 140 mm)Compartment Dimensions: 5 3/8 x 2 1/16 x 5 1/4inches (137 x 51.6 x 133mm)
Catalog No. 50-302-7997 Supplier Alkali Scientific Supplier No. UFDR362S
For Standard 2″ BoxesRack Only Upright freezer racks for storage of 2 inch boxes, features a 5 boxes deep by 5 boxes high configuration for a total storage of 25 boxes per rack. These upright freezer drawer racks are built with durable, corrosion resistant stainless steel. Each rack includes separate drawers with handles allowing for an easy retrieval from a laboratory freezer without the need to pull out the entire rack. This drawer style freezer rack does not come standard with storage boxes. This rack is compatible with the PHCbi VIP freezers. Please see theFreezer Rack Configurator if you are unsure of what rack you may need. Dimensions (D x H x W): 16 7/8 x 12 5/8 x 5 1/2  inches (428 x 321 x 140 mm)Compartment Dimensions: 5 3/8 x 2 1/16 x 5 1/4inches (137 x 51.6 x 133mm)