Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack with Drawers for Standard 3″ Boxes; Top-Loading Rack; Rack Only; 4x4 Configuration, 16 Box Capacity
SDP
Catalog No. 503028051
Encompass_Preferred
For Standard 3″ BoxesRack Only – Top-loaded drawer rack Upright Freezer Drawer Racks are for Standard 3" fiberboard boxes.  Racks are constructed of corrosion resistant stainless steel, including handles, identification card holders and have a top-loading system to insert boxes.  Upright Freezer Drawer racks offer an organized system for storage and retrieval of samples.  Drawer racks facilitate both access to and storage of samples without having to remove the whole rack from the freezer.  Security lock devices are also available for each rack though ordered separately. 22 3/8 x 16 9/16 x 5 ½ inch (566 x 420 x 140 mm)5 1/2 x 3 1/8 x 5 ¼ inch (137.4 x 79.4 x 133 mm)
Catalog No. 50-302-8051 Supplier Alkali Scientific Supplier No. UFDR443S
For Standard 3″ BoxesRack Only – Top-loaded drawer rack Upright Freezer Drawer Racks are for Standard 3" fiberboard boxes.  Racks are constructed of corrosion resistant stainless steel, including handles, identification card holders and have a top-loading system to insert boxes.  Upright Freezer Drawer racks offer an organized system for storage and retrieval of samples.  Drawer racks facilitate both access to and storage of samples without having to remove the whole rack from the freezer.  Security lock devices are also available for each rack though ordered separately. 22 3/8 x 16 9/16 x 5 ½ inch (566 x 420 x 140 mm)5 1/2 x 3 1/8 x 5 ¼ inch (137.4 x 79.4 x 133 mm)