Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Rack with Drawers for Standard 2″ Boxes; Top-Loading Rack; Rack Only; 4x5 Configuration, 20 Box Capacity
SDP
Catalog No. 503028026
Encompass_Preferred
These Upright Freezer Drawer Racks are constructed of corrosion resistant stainless steel, including handles, identification card holders and have a top-loading system to insert boxes.  Upright Freezer Drawer racks offer a more organized system for storage and retrieval of samples.  Drawer racks facilitate both access to and storage of samples without having to remove the whole rack from the freezer. Dimensions: 22 5/16 x 11 x 5 ½ inch (566 x 278.6 x 140 mm)Compartment Size 5 7/16 x 2 3/16 x 5 3/16 inch (137.5 x 54.5 x 134.5 mm)
Catalog No. 50-302-8026 Supplier Alkali Scientific Supplier No. UFDR452ST
These Upright Freezer Drawer Racks are constructed of corrosion resistant stainless steel, including handles, identification card holders and have a top-loading system to insert boxes.  Upright Freezer Drawer racks offer a more organized system for storage and retrieval of samples.  Drawer racks facilitate both access to and storage of samples without having to remove the whole rack from the freezer. Dimensions: 22 5/16 x 11 x 5 ½ inch (566 x 278.6 x 140 mm)Compartment Size 5 7/16 x 2 3/16 x 5 3/16 inch (137.5 x 54.5 x 134.5 mm)