Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks, 100-Cell Hinged Top Plastic Boxes, (4x3) 16 Box Capacity
SDP
Catalog No. 503026693
Encompass_Preferred
For 100 Cell Hinged Lid Storage Constructed of corrosion resistant stainless steel Racks are for 100-Cell Hinged Top Plastic Boxes Security Lock devices are available. Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right- or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Racks are for 100-Cell Hinged Top Plastic Boxes. Constructed of corrosion resistant stainless steel. Security Lock devices are available for each rack if required. Dimensions l x h x w: 23 1/16 x 9 3/4 x 6 7/16 inches (442 x 247 x 164 mm)Compartment Sizes: 5 5/8 x 2 3/16 x 6 1/8 inches (142.8 x 56 x 156mm) *Lock device is ordered separately.
Catalog No. 50-302-6693 Supplier Alkali Scientific Supplier No. UFDRHT44
For 100 Cell Hinged Lid Storage Constructed of corrosion resistant stainless steel Racks are for 100-Cell Hinged Top Plastic Boxes Security Lock devices are available. Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right- or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Racks are for 100-Cell Hinged Top Plastic Boxes. Constructed of corrosion resistant stainless steel. Security Lock devices are available for each rack if required. Dimensions l x h x w: 23 1/16 x 9 3/4 x 6 7/16 inches (442 x 247 x 164 mm)Compartment Sizes: 5 5/8 x 2 3/16 x 6 1/8 inches (142.8 x 56 x 156mm) *Lock device is ordered separately.