Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks, 100-Cell Hinged Top Plastic Boxes, 6 Box Capacity
SDP
Catalog No. 503029103
Encompass_Preferred
For 100 Cell Hinged Lid Storage 2×3 Configuration Constructed of corrosion resistant stainless steel Racks are for 100-Cell Hinged Top Plastic Boxes Security Lock devices are available. Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right- or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Racks are for 100-Cell Hinged Top Plastic Boxes. Constructed of corrosion resistant stainless steel. Security Lock devices are available for each rack if required.   Dimensions l x h x w (11 3/4 x 7 5/16 x 6 7/16 Inches ( 299 x 56 x 156mm)Compartment Dimensions: 5 5/8 x 6 1/8 x 2 3/16 inches (142.8 x 156 x 56mm) *Lock device is ordered separately.
Catalog No. 50-302-9103 Supplier Alkali Scientific Supplier No. UFDRHT23
For 100 Cell Hinged Lid Storage 2×3 Configuration Constructed of corrosion resistant stainless steel Racks are for 100-Cell Hinged Top Plastic Boxes Security Lock devices are available. Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right- or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Racks are for 100-Cell Hinged Top Plastic Boxes. Constructed of corrosion resistant stainless steel. Security Lock devices are available for each rack if required.   Dimensions l x h x w (11 3/4 x 7 5/16 x 6 7/16 Inches ( 299 x 56 x 156mm)Compartment Dimensions: 5 5/8 x 6 1/8 x 2 3/16 inches (142.8 x 156 x 56mm) *Lock device is ordered separately.