Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks, 100-Cell Hinged Top Plastic Boxes, 2 x 6 Configuration, 12 Box Capacity
SDP
Catalog No. 503028072
Encompass_Preferred
For 100 Cell Hinged Lid Storage Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right- or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Racks are for 100-Cell Hinged Top Plastic Boxes. Constructed of corrosion resistant stainless steel. Security Lock devices are available for each rack if required. Constructed of corrosion resistant stainless steel Racks are for 100-Cell Hinged Top Plastic Boxes Security Lock devices are available. Dimensions l x h x w: 11 3/4 x 14 1/2 x 6 7/16 inches (299 x 369 x 164 mm)Compartment Sizes: 5 5/8 x 2 3/16 x 6 1/8 inches (142.8 x 56 x 156mm) *Lock device is ordered separately.
Catalog No. 50-302-8072 Supplier Alkali Scientific Supplier No. UFDRHT26
For 100 Cell Hinged Lid Storage Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right- or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Racks are for 100-Cell Hinged Top Plastic Boxes. Constructed of corrosion resistant stainless steel. Security Lock devices are available for each rack if required. Constructed of corrosion resistant stainless steel Racks are for 100-Cell Hinged Top Plastic Boxes Security Lock devices are available. Dimensions l x h x w: 11 3/4 x 14 1/2 x 6 7/16 inches (299 x 369 x 164 mm)Compartment Sizes: 5 5/8 x 2 3/16 x 6 1/8 inches (142.8 x 56 x 156mm) *Lock device is ordered separately.