Learn More
Thomas Scientific Upright Freezer Racks for 6 x 100 Place Hinged Top Boxes, Holds 6 Boxes (2x3), 12 1/2"L x 6 15/16"H x 5 7/8"D
SDP
Catalog No. 50195975
Encompass_Preferred
Racks are for 100-Cell Hinged Top Plastic Boxes. Constructed of corrosion resistant stainsteel.
Catalog No. 50-195-975 Supplier Thomas Scientific Supplier No. 1148X80
Racks are for 100-Cell Hinged Top Plastic Boxes. Constructed of corrosion resistant stainsteel.