Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for 100-Cell Hinged Plastic Storage Boxes, Holds 15 Boxes
SDP
Catalog No. 503027864
Encompass_Preferred
For Large 2″ Cardboard Boxes and 100-Cell Hinged Top Plastic Storage Boxes Upright freezer racks for storage of 100-cell hinged top boxes, features 3 boxes deep by 5 boxes high configuration for a total of 15 boxes. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. Dimensions (DxWxH): 19 3/16 x 5 7/8 x 11 9/16 inches (488 x 149 x 293mm)Compartment Dimensions: 6 1/8 x 5 13/16 x 2 5/16 inches (156 x 147.5 x 57.6mm) *Lock device is ordered separately
Catalog No. 50-302-7864 Supplier Alkali Scientific Supplier No. UFHT35
For Large 2″ Cardboard Boxes and 100-Cell Hinged Top Plastic Storage Boxes Upright freezer racks for storage of 100-cell hinged top boxes, features 3 boxes deep by 5 boxes high configuration for a total of 15 boxes. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. Dimensions (DxWxH): 19 3/16 x 5 7/8 x 11 9/16 inches (488 x 149 x 293mm)Compartment Dimensions: 6 1/8 x 5 13/16 x 2 5/16 inches (156 x 147.5 x 57.6mm) *Lock device is ordered separately