Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for 100-Cell Hinged Plastic Storage Boxes, Holds 8 Boxes
SDP
Catalog No. 503028537
Encompass_Preferred
For Large 2″ Cardboard Boxes and 100-Cell Hinged Top Plastic Storage Boxes Upright freezer racks for storage of 100-cell hinged top boxes, features 2 boxes deep by 4 boxes high configuration for a total of 10 boxes. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. Dimensions (DxWxH): 13 x 5 7/8 x 9 1/4 inches (330 x 149 x 235mm)Compartment Dimensions: 6 1/8 x 5 13/16 x 2 5/16 inches (156 x 147.5 x 57.6mm) *Lock device is ordered separately.
Catalog No. 50-302-8537 Supplier Alkali Scientific Supplier No. UFHT24
For Large 2″ Cardboard Boxes and 100-Cell Hinged Top Plastic Storage Boxes Upright freezer racks for storage of 100-cell hinged top boxes, features 2 boxes deep by 4 boxes high configuration for a total of 10 boxes. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. Dimensions (DxWxH): 13 x 5 7/8 x 9 1/4 inches (330 x 149 x 235mm)Compartment Dimensions: 6 1/8 x 5 13/16 x 2 5/16 inches (156 x 147.5 x 57.6mm) *Lock device is ordered separately.