Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for 100-Cell Hinged Plastic Storage Boxes, Holds 12 Boxes (4x3)
SDP
Catalog No. 503028077
Encompass_Preferred
For Large 2″ Cardboard Boxes and 100-Cell Hinged Top Plastic Storage Boxes Upright freezer racks for storage of 100-cell hinged top boxes, features 4 boxes deep by 3 boxes high configuration for a total of 12 boxes. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. Dimensions (DxWxH): 25 7/16 x 5 7/8 x 6 15/16 inches (646 x 149 x 176mm)Compartment Dimensions: 6 1/8 x 5 13/16 x 2 5/16 inches (156 x 147.5 x 57.6mm) *Lock device is ordered separately.
Catalog No. 50-302-8077 Supplier Alkali Scientific Supplier No. UFHT43
For Large 2″ Cardboard Boxes and 100-Cell Hinged Top Plastic Storage Boxes Upright freezer racks for storage of 100-cell hinged top boxes, features 4 boxes deep by 3 boxes high configuration for a total of 12 boxes. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. Dimensions (DxWxH): 25 7/16 x 5 7/8 x 6 15/16 inches (646 x 149 x 176mm)Compartment Dimensions: 6 1/8 x 5 13/16 x 2 5/16 inches (156 x 147.5 x 57.6mm) *Lock device is ordered separately.