Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for 100-Cell Hinged Plastic Storage Boxes, 3x6, Holds 18 Boxes
SDP
Catalog No. 503028561
Encompass_Preferred
For Large 2″ Cardboard Boxes and 100-Cell Hinged Top Plastic Storage Boxes Upright freezer racks for storage of 100-cell hinged top boxes, features 3 boxes deep by 6 boxes high configuration for a total of 18 boxes. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. Dimensions (DxWxH): 19 3/16 x 5 7/8 x 13 13/16 inches (488 x 149 x 351mm)Compartment Dimensions: 6 1/8 x 5 13/16 x 2 5/16 inches (156 x 147.5 x 57.6mm)
Catalog No. 50-302-8561 Supplier Alkali Scientific Supplier No. UFHT36
For Large 2″ Cardboard Boxes and 100-Cell Hinged Top Plastic Storage Boxes Upright freezer racks for storage of 100-cell hinged top boxes, features 3 boxes deep by 6 boxes high configuration for a total of 18 boxes. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. Dimensions (DxWxH): 19 3/16 x 5 7/8 x 13 13/16 inches (488 x 149 x 351mm)Compartment Dimensions: 6 1/8 x 5 13/16 x 2 5/16 inches (156 x 147.5 x 57.6mm)