Learn More
Thomas Scientific Freezer Rack for 2" boxes, 3 x 4 cap
SDP
Catalog No. 50195990
Encompass_Preferred
Stainless Steel Standard Freezer Rack for Upright Freezers for use with 2" or 3" Boxes
Catalog No. 50-195-990 Supplier Thomas Scientific Supplier No. 1201X90
Stainless Steel Standard Freezer Rack for Upright Freezers for use with 2" or 3" Boxes