Learn More
Thomas Scientific Freezer Rack for 2" boxes, 4 x 4 cap
SDP
Catalog No. 50196005
Encompass_Preferred
Stainless Steel Standard Freezer Rack for Upright Freezers for use with 2" or 3" Boxes
Catalog No. 50-196-005 Supplier Thomas Scientific Supplier No. 1201X95
Stainless Steel Standard Freezer Rack for Upright Freezers for use with 2" or 3" Boxes