Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for 3.75″ Boxes- Rack Only- 2 Boxes Deep x 2 Boxes High
SDP
Catalog No. 503028087
Encompass_Preferred
Standard upright freezer racks for storage of 3.75-inch boxes, feature 2 boxes deep by 2 boxes high configuration for a total storage of 4 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. This freezer rack does not come standard with storage boxes. Dimensions L x H x W: 12 7/16 x 8 1/16 x 5 15/16 (315.5 x 205 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)
Catalog No. 50-302-8087 Supplier Alkali Scientific Supplier No. UFR22L375
Standard upright freezer racks for storage of 3.75-inch boxes, feature 2 boxes deep by 2 boxes high configuration for a total storage of 4 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for easy retrieval from a laboratory freezer. This freezer rack does not come standard with storage boxes. Dimensions L x H x W: 12 7/16 x 8 1/16 x 5 15/16 (315.5 x 205 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)