Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for 3.75″ Boxes- Rack Only- 3 Boxes Deep x 3 Boxes High
SDP
Catalog No. 503028090
Encompass_Preferred
Standard upright freezer racks for storage of 3.75-inch boxes, features a 3 boxes deep by 3 boxes high configuration for a total storage of 9 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for an easy retrieval from a laboratory freezer. This freezer rackdoes not come standard with storage boxes. Dimensions L x H x W: 16 13/16 x 12 1/16 x 5 1/2 inch (427 x 307 x 140mm)Compartment Size L x H x W: 5 3/8 x 3 15/16 x 5 3/8 inch (136 x 100 x137mm)
Catalog No. 50-302-8090 Supplier Alkali Scientific Supplier No. UFR33375
Standard upright freezer racks for storage of 3.75-inch boxes, features a 3 boxes deep by 3 boxes high configuration for a total storage of 9 boxes per rack. These upright freezer racks are built with durable, corrosion resistant stainless steel. Each rack includes handles for an easy retrieval from a laboratory freezer. This freezer rackdoes not come standard with storage boxes. Dimensions L x H x W: 16 13/16 x 12 1/16 x 5 1/2 inch (427 x 307 x 140mm)Compartment Size L x H x W: 5 3/8 x 3 15/16 x 5 3/8 inch (136 x 100 x137mm)