Learn More
Thomas Scientific Upright Freezer Rack for 15 & 50mL Tube Boxes, 3 x 2
SDP
Catalog No. 50195998
Encompass_Preferred
Thomas No. 1180Q43 Box Capacity: 2 x 2 = 4 Shipping Weight: 1.67 kg / 3.68 lb Outside Dimension: inch (mm) L: 12 3 /16 (310) H: 10 3 /16 (259) W: 5 7 /8 (149) Lock Device Order Separately: DP-1210 Description: Rack for 15 ml and 50 ml Tube Boxes Thomas No 1180Q44 Box Capacity : 3 x 2 = 6 Shipping Weight: 2.36 kg / 5.2 lb Outside Dimension: inch (mm) L: 18 1 /4 (464) H: 10 3 /16 (259) W: 5 7 /8 (149) Lock Device Order Separately: DP-1810 Description: Rack for 15 ml and 50 ml Tube Boxes Thomas No.1180Q45 Box Capacity : 4 x 2 = 8 Shipping Weight: 2.98 kg / 6.57 lb Outside Dimension: inch (mm) L: 24 5 /16 (617) H: 10 3 /16 (259) W: 5 7 /8 (149) Lock Device Order Separately: DP-2410 Description: Rack for 15 ml and 50 ml Tube Boxes+C7
Catalog No. 50-195-998 Supplier Thomas Scientific Supplier No. 1180Q44
Thomas No. 1180Q43 Box Capacity: 2 x 2 = 4 Shipping Weight: 1.67 kg / 3.68 lb Outside Dimension: inch (mm) L: 12 3 /16 (310) H: 10 3 /16 (259) W: 5 7 /8 (149) Lock Device Order Separately: DP-1210 Description: Rack for 15 ml and 50 ml Tube Boxes Thomas No 1180Q44 Box Capacity : 3 x 2 = 6 Shipping Weight: 2.36 kg / 5.2 lb Outside Dimension: inch (mm) L: 18 1 /4 (464) H: 10 3 /16 (259) W: 5 7 /8 (149) Lock Device Order Separately: DP-1810 Description: Rack for 15 ml and 50 ml Tube Boxes Thomas No.1180Q45 Box Capacity : 4 x 2 = 8 Shipping Weight: 2.98 kg / 6.57 lb Outside Dimension: inch (mm) L: 24 5 /16 (617) H: 10 3 /16 (259) W: 5 7 /8 (149) Lock Device Order Separately: DP-2410 Description: Rack for 15 ml and 50 ml Tube Boxes+C7