Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks, for 60 Well & 384 Well Microtiter Plates, Holds 60 with Lids, 80 without Lids
SDP
Catalog No. 503028507
Encompass_Preferred
For Speciality Tube Racks (Micronic,Fluidx, Abgene )- H: 1 1/4″-1 1/2″(31.7-38.7mm) Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Durable stainless steel freezer racks facilitate organization and maximize valuable storage space..  Handles at both ends accommodate freezers with right or left handed doors.  Racks are for 96-well & 384-well Microtiter Plates.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Features: Racks are for 96-well & 384-well Microtiter Plates Constructed of corrosion resistant stainless steel Sturdy handle design Dimensions L x H x W: 17 7/8 x 7 3/8 x 5 7/16inch (454 x 188 x 138mm)Compartment l x h x w: 3 7/16 x 1 5/8 x 5 1/8 inch (88 x 41.4 x 130mm)
Catalog No. 50-302-8507 Supplier Alkali Scientific Supplier No. UFDRML54
For Speciality Tube Racks (Micronic,Fluidx, Abgene )- H: 1 1/4″-1 1/2″(31.7-38.7mm) Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Durable stainless steel freezer racks facilitate organization and maximize valuable storage space..  Handles at both ends accommodate freezers with right or left handed doors.  Racks are for 96-well & 384-well Microtiter Plates.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers. Features: Racks are for 96-well & 384-well Microtiter Plates Constructed of corrosion resistant stainless steel Sturdy handle design Dimensions L x H x W: 17 7/8 x 7 3/8 x 5 7/16inch (454 x 188 x 138mm)Compartment l x h x w: 3 7/16 x 1 5/8 x 5 1/8 inch (88 x 41.4 x 130mm)