Learn More
Thomas Scientific Upright Freezer Racks for 8 x 100 Place Hinged Top Boxes, Holds 8 Boxes (4x2), 12 1/2"L x 9 1/4"H x 5 7/8"D
SDP
Catalog No. 50195979
Encompass_Preferred
Racks are for 100-Cell Hinged Top Plastic Boxes. Constructed of corrosion resistant stainsteel.
Catalog No. 50-195-979 Supplier Thomas Scientific Supplier No. 1148X74
Racks are for 100-Cell Hinged Top Plastic Boxes. Constructed of corrosion resistant stainsteel.