Learn More
Thomas Scientific Rack Microplates 22 x 11 x 5 1/2
SDP
Catalog No. 50196008
Encompass
Racks are for 96-well & 384-well Microtiter Plates. Constructed of corrosion resistant stainsteel.
Catalog No. 50-196-008 Supplier Thomas Scientific Supplier No. 1149P28
Racks are for 96-well & 384-well Microtiter Plates. Constructed of corrosion resistant stainsteel.