Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for 96 Deep-Well Microtiter Plates and Microtube Racks with Locking Rod – Rack Only – 6 x 5, 30 Box Capacity
SDP
Catalog No. 503029480
Encompass_Preferred
For 96 Deep-Well Microtiter PlatesFor Speciality Tube Racks (Matrix, Fluidx, Micronic)- H: 9/16″- 1 3/4″ (14.4-44.32mm) 6×5 Configuration Upright freezer racks are designed for storage of 96 Deep-Well Microtiter Plates and Microtube Racks. This freezer rack features a 6 high by 5 deep configuration for a total storage of 30 plates or microtube racks. These high-quality upright freezer racks include handles, are constructed of corrosion resistant stainless steel and offer convenient storage of samples in an organized format. Each rack compartment holds a single 96 deep-well microtiter plate or microtube rack/box. A locking rod is included with this freezer rack to provide added security to prevent plates or boxes from sliding out of the rack when moving the racks in and out of the freezer. Dimensions (DxWxH): 22 1/2 x 5 1/2 x 9 5/16 inches (571 x 140 x 237mm)Compartment Dimensions: 3 9/16 x 5 3/16 x 1 13/16 inches (91 x 130.5 x 45.7mm)
Catalog No. 50-302-9480 Supplier Alkali Scientific Supplier No. UFDP65L
For 96 Deep-Well Microtiter PlatesFor Speciality Tube Racks (Matrix, Fluidx, Micronic)- H: 9/16″- 1 3/4″ (14.4-44.32mm) 6×5 Configuration Upright freezer racks are designed for storage of 96 Deep-Well Microtiter Plates and Microtube Racks. This freezer rack features a 6 high by 5 deep configuration for a total storage of 30 plates or microtube racks. These high-quality upright freezer racks include handles, are constructed of corrosion resistant stainless steel and offer convenient storage of samples in an organized format. Each rack compartment holds a single 96 deep-well microtiter plate or microtube rack/box. A locking rod is included with this freezer rack to provide added security to prevent plates or boxes from sliding out of the rack when moving the racks in and out of the freezer. Dimensions (DxWxH): 22 1/2 x 5 1/2 x 9 5/16 inches (571 x 140 x 237mm)Compartment Dimensions: 3 9/16 x 5 3/16 x 1 13/16 inches (91 x 130.5 x 45.7mm)