Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for 96-Well and 384-Well Microtiter Plates with Locking Rod – Rack Only (98-112 Plates)
SDP
Catalog No. 503027601
Encompass_Preferred
96-Well and 384-Well Microtiter Plates Upright Freezer Racks are designed for storage of 96 & 384-Well Microtiter Plates and can store up to 112 plates. These freezer racks are constructed of corrosion resistant stainless steel, include handles, and offer convenient storage of microtiter plates in your laboratory freezer.  Locking rods are also included for added security. Microtiter plates are not included with purchase. Plate Capacity: 98 with lids 112 without lids Dimensions (DxWxH): 26 1/8 x 5 1/2 x 9 3/16 inches (571 x 140 x 233mm)Compartment Dimensions: 3 9/16 x 5 3/16 x 4 1/2 inches (91 x 130.5 x 115mm) Lock device is ordered separately.
Catalog No. 50-302-7601 Supplier Alkali Scientific Supplier No. UFRMP709L
96-Well and 384-Well Microtiter Plates Upright Freezer Racks are designed for storage of 96 & 384-Well Microtiter Plates and can store up to 112 plates. These freezer racks are constructed of corrosion resistant stainless steel, include handles, and offer convenient storage of microtiter plates in your laboratory freezer.  Locking rods are also included for added security. Microtiter plates are not included with purchase. Plate Capacity: 98 with lids 112 without lids Dimensions (DxWxH): 26 1/8 x 5 1/2 x 9 3/16 inches (571 x 140 x 233mm)Compartment Dimensions: 3 9/16 x 5 3/16 x 4 1/2 inches (91 x 130.5 x 115mm) Lock device is ordered separately.