Learn More
Labrepco Inc UPRIGHT FREEZER RACKS FOR 96-W
SDP
Catalog No. 509020264
Encompass_Preferred
Rack, Upright freezer; Labrepco; Crystal; For 96-Well and 384-Well Microtiter Plates; Capacity: 84 Plates w/lids, 96 Plates w/o lids; L x H x D: 559 x 233 x 140mm
Catalog No. 50-902-0264 Supplier Labrepco Inc Supplier No. UFMP609L
Rack, Upright freezer; Labrepco; Crystal; For 96-Well and 384-Well Microtiter Plates; Capacity: 84 Plates w/lids, 96 Plates w/o lids; L x H x D: 559 x 233 x 140mm