Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for 96-Well and 384-Well Plates with Locking Rod – Rack Only (42-48 Plates)
SDP
Catalog No. 503027811
Encompass_Preferred
For 96-well & 384- Well Microtiter Plates Upright Freezer Racks are designed for storage of 96 & 384-Well Microtiter Plates and can store up to 80 plates. These freezer racks are constructed of corrosion resistant stainless steel, include handles, and offer convenient storage of microtiter plates in your laboratory freezer.  Locking rods are also included for added security. Microtiter plates are not included with purchase. Plate Capacity: 42 with lids 48 without lids Dimensions (DxWxH): 11 1/2 x 5 1/2 x 9 3/16 inches (292 x 140 x 233mm)Compartment Dimensions: 3 9/16 x 5 3/16 x 4 1/2 inches (91 x 130.5 x 115mm)
Catalog No. 50-302-7811 Supplier Alkali Scientific Supplier No. UFRMP309L
For 96-well & 384- Well Microtiter Plates Upright Freezer Racks are designed for storage of 96 & 384-Well Microtiter Plates and can store up to 80 plates. These freezer racks are constructed of corrosion resistant stainless steel, include handles, and offer convenient storage of microtiter plates in your laboratory freezer.  Locking rods are also included for added security. Microtiter plates are not included with purchase. Plate Capacity: 42 with lids 48 without lids Dimensions (DxWxH): 11 1/2 x 5 1/2 x 9 3/16 inches (292 x 140 x 233mm)Compartment Dimensions: 3 9/16 x 5 3/16 x 4 1/2 inches (91 x 130.5 x 115mm)