Learn More
Labrepco Inc UPRIGHT FREEZER RACKS FOR 96-W
SDP
Supplier:  Labrepco Inc UFMP509L
Encompass_Preferred
Rack, Upright freezer; Labrepco; Crystal; For 96-Well and 384-Well Microtiter Plates; Capacity: 70 Plates w/lids, 80 Plates w/o lids; L x H x D: 466 x 233 x 140mm
Catalog No. 50-902-0262