Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for Large 3.75″ Boxes- Rack Only- 3 Boxes Deep x 3 Boxes High
SDP
Catalog No. 503026396
Encompass_Preferred
These Upright Freezer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for easy removal from freezer. Dimensions L x H x W: 18 3/8 x 12 1/16 x 5 15/16 (315.5 x 307 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)
Catalog No. 50-302-6396 Supplier Alkali Scientific Supplier No. UFR33L375
These Upright Freezer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for easy removal from freezer. Dimensions L x H x W: 18 3/8 x 12 1/16 x 5 15/16 (315.5 x 307 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)