Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for Large 3.75″ Boxes- Rack Only- 2 Boxes Deep x 4 Boxes High
SDP
Catalog No. 503027906
Encompass_Preferred
These Upright Freezer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for an easy. Dimensions L x H x W: 12 7/16 x 16 1/126 x 5 15/16 (315.5 x 408 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm
Catalog No. 50-302-7906 Supplier Alkali Scientific Supplier No. UFR24L375
These Upright Freezer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for an easy. Dimensions L x H x W: 12 7/16 x 16 1/126 x 5 15/16 (315.5 x 408 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm