Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for Large 3.75″ Boxes- Rack Only- 3 Boxes Deep x 2 Boxes High
SDP
Catalog No. 503028102
Encompass_Preferred
These Upright Freezer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for easy removal from freezer. Dimensions L x H x W: 18 3/8 x 8 1/16 x 5 15/16 (315.5 x 205 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)
Catalog No. 50-302-8102 Supplier Alkali Scientific Supplier No. UFR32L375
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These Upright Freezer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for easy removal from freezer. Dimensions L x H x W: 18 3/8 x 8 1/16 x 5 15/16 (315.5 x 205 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)