Learn More
Alkali Scientific Alkali Scientific™ Upright Freezer Racks for Large 3.75″ Boxes- Rack Only- 2 Boxes Deep x 3 Boxes High
SDP
Catalog No. 503027734
Encompass_Preferred
These Upright Freezer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for easy removal from freezer. Dimensions L x H x W: 12 7/16 x 12 1/16  x 5 15/16 (315.5 x 307 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)
Catalog No. 50-302-7734 Supplier Alkali Scientific Supplier No. UFR23L375
These Upright Freezer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for easy removal from freezer. Dimensions L x H x W: 12 7/16 x 12 1/16  x 5 15/16 (315.5 x 307 x 151mm)Compartment Dimensions: 5 7/8 x 4 x 5 13/16 inch (149 x 101 x 148mm)