Learn More
Labrepco Inc UPRIGHT FREEZER RACKS FOR STD
SDP
Supplier:  Labrepco Inc UF542
Encompass_Preferred
Rack, Upright freezer; Labrepco; Crystal; For standard 2 in. boxes; Capacity (Box): 20; L x H x D: 679 x 225 x 140mm
Catalog No. 50-902-0102