Learn More
Labrepco Inc Upright Freezer Rack with Drawers for Hinged Top Plastic Boxes- Rack Only- 4 Deep x 3 High
SDP
Catalog No. NC1111327
Encompass_Preferred
Stainless Steel Rack with 3 drawers (4 compartments per drawer) for twelve 100-Cell Hinged Top Plastic Storage Boxes. Boxes and cell dividers not included. Drawer style rack design allows easy access to boxes thus protecting other samples in the rack from warming up.
Catalog No. NC1111327 Supplier Labrepco Inc Supplier No. UFDHT43
Stainless Steel Rack with 3 drawers (4 compartments per drawer) for twelve 100-Cell Hinged Top Plastic Storage Boxes. Boxes and cell dividers not included. Drawer style rack design allows easy access to boxes thus protecting other samples in the rack from warming up.