Learn More
Yamato Scientific America, Inc. Yamato Upright Style Low Temperature Freezer 0ºc To -40ºc, 22 Cu.Ft., Manual Defrost
Catalog No. NC1869648 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Low temperature upright style freezers comes standard with microprocessor digital controller, audible and visual alarms, remote alarm contacts, alarm battery back-up, and 1/2" access port.
Catalog No. NC1869648 Supplier Yamato Scientific America, Inc. Supplier No. LTF601U
Add to Cart
Add to Cart
Request a Quote
Low temperature upright style freezers comes standard with microprocessor digital controller, audible and visual alarms, remote alarm contacts, alarm battery back-up, and 1/2" access port.