Learn More
Felix Storch Inc ACCUCOLD Upright Pharmacy Freezer Capacity with Probe hole
SDP
Supplier:  Felix Storch Inc AFS23MLLHPHRIL
Encompass_Preferred
The AFS23MLLHPRHRL is a 23 cu.ft. all-freezer with an adjustable control range from -30 to -15C. The microprocessor temperature controller features a large digital LED display viewable in Celsius or Fahrenheit, making it easy for staff to manage and monitor the temperature. The unit is equipped with an air sensor for rapid control response and a product sensor for a more accurate reading of the interior, including a display of the min/max temperature in 15 minute increments The audible and visual temperature alarm sounds if the unit rises or falls above a user-set range. Additional alarms include a power failure alert, door open alarm, and a condenser cleaning reminder. Probe Hole (installed on left side)Features a durable stainless steel cabinet and interior, with a sealed back for easier cleanup. A factory-installed lock with two keys included. Magnetic door gaskets ensure a strong seal. Door is self-closing to help protect contents, but will remain open at 90 for easier loading.
Catalog No. NC3901294
Only null left
Add to Cart
Request a Quote