Learn More
Felix Storch Inc ACCUCOLD Upright Pharmacy Refrigerator 23 Cu.Ft. Capacity with Probe hole
SDP
Catalog No. NC3901262
Encompass_Preferred
The ARG23MLPHRIR is a 23 cu.ft. glass door all-refrigerator with an adjustable control range from +2 to +10C. The microprocessor temperature controller features a large digital LED display viewable in Celsius or Fahrenheit, making it easy for staff to manage and monitor the temperature. The unit is equipped with an air sensor for rapid control response and a product sensor for a more accurate reading of the interior, including a display of the min/max temperature in 15 minute increments The audible and visual temperature alarm sounds if the unit rises or falls above a user-set range. Additional alarms include a power failure alert, door open alarm, and a condenser cleaning reminder. Probe Hole (installed on right side)Features a durable stainless steel cabinet and interior, with a sealed back for easier cleanup. The double pane tempered glass door provides a full display of the interior. A factory-installed lock with two keys included for your facility.
Catalog No. NC3901262 Supplier Felix Storch Inc Supplier No. ARG23MLPHRIR
Add to Cart
Add to Cart
Request a Quote
The ARG23MLPHRIR is a 23 cu.ft. glass door all-refrigerator with an adjustable control range from +2 to +10C. The microprocessor temperature controller features a large digital LED display viewable in Celsius or Fahrenheit, making it easy for staff to manage and monitor the temperature. The unit is equipped with an air sensor for rapid control response and a product sensor for a more accurate reading of the interior, including a display of the min/max temperature in 15 minute increments The audible and visual temperature alarm sounds if the unit rises or falls above a user-set range. Additional alarms include a power failure alert, door open alarm, and a condenser cleaning reminder. Probe Hole (installed on right side)Features a durable stainless steel cabinet and interior, with a sealed back for easier cleanup. The double pane tempered glass door provides a full display of the interior. A factory-installed lock with two keys included for your facility.