Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Side Access Freezer Rack, Holds (36) 96 Well PCR Boxes,Rack Dims 22 1/2L x 9 1/8H x5 1/2W" (571 x 231 x 140mm), Compartment Dims 3 1/2L x 1 7/16H x 5 3/16W" (91 x 37 x 130mm) 6x6
Catalog No. NC2208111
Encompass_Preferred
Rack Designed to hold (36) 0.2ml Tube 96-Well PCR Boxes (1 5/16H x 5 1/32W x 3 1/2L" (34 x 127 x 89mm), Locking Rods Included, made of corrosion resistant stainless steel, for use in Freezers or LN2 Tanks, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available.
This item is not returnable. View return policy
Catalog No. NC2208111 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFPCR66L
Rack Designed to hold (36) 0.2ml Tube 96-Well PCR Boxes (1 5/16H x 5 1/32W x 3 1/2L" (34 x 127 x 89mm), Locking Rods Included, made of corrosion resistant stainless steel, for use in Freezers or LN2 Tanks, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available.